Learn More
MSA™ Comfo-Cap™ Protective Cap
Catalog No. 19088095 Shop All MSA Products
Change view
Click to view available options
Color:
Black
Green
Red
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-088-095 Black
19-088-096 Green
19-088-097 Red
19-088-714 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-088-095 Supplier MSA™ Supplier No. 82769
Add to Cart
Add to Cart
Low profile makes this cap ideal for mining applications
  • Low-crown cap, designed for work in low-ceiling areas
  • Includes lamp bracket and cordholder to accommodate a miner's lamp
  • Comfortable, lightweight, polycarbonate plastic shell resists top impact
  • Molded cross ridges on crown provide increased rigidity and strength
  • Peak helps prevent debris from getting near face
  • Staz-On™ pinlock and Fas-Trac ratchet suspension options available
Compliance
Third-party certified by SEI to meet ANSI/ISEA Z89.1, Type I, Class G requirements.
Specifications
Certifications/Compliance Meets or exceeds ANSI Z89.1-1997, Class G (electrical-low voltage).
Color Black
Material Polycarbonate
Size Standard
Suspension Type Pin-Lock
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.