Learn More
MSA™ Comfo-Cap™ Protective Cap
Catalog No. 19088097 Shop All MSA Products
Change view
Click to view available options
Color:
Black
Green
Red
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-088-097 Red
19-088-096 Green
19-088-714 White
19-088-095 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-088-097 Supplier MSA™ Supplier No. 91590
Add to Cart
Add to Cart
Low profile makes this cap ideal for mining applications
  • Low-crown cap, designed for work in low-ceiling areas
  • Includes lamp bracket and cordholder to accommodate a miner's lamp
  • Comfortable, lightweight, polycarbonate plastic shell resists top impact
  • Molded cross ridges on crown provide increased rigidity and strength
  • Peak helps prevent debris from getting near face
  • Staz-On™ pinlock and Fas-Trac ratchet suspension options available
Compliance
Third-party certified by SEI to meet ANSI/ISEA Z89.1, Type I, Class G requirements.
Specifications
Certifications/Compliance Meets or exceeds ANSI Z89.1-1997, Class G (electrical-low voltage).
Color Red
Material Polycarbonate
Size Standard
Suspension Type Pin-Lock