Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Comfo™ Cartridges

Catalog No. 19003216 Shop All MSA Products
Change view
Click to view available options
Filter Type:
N95, R951
P100
Cartridge Type:
Ammonia, Methylamine Cartridge
Chlorine, Mercury Vapor Cartridge
Organic Vapor Cartridge
Organic Vapor, Acid Gas Cartridge
Organic Vapor, Acid Gas, Ammonia, Methylamine, Formaldehyde, Hydrogen Fluoride Cartridge
Particulate Cartridge
Design Type:
Low-profile
Short Stack
Standard
Quantity:
10/Pk.
6/Pk.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type Cartridge Type Design Type Quantity
19-003-216 P100 Organic Vapor Cartridge Standard 6/Pk.
18-921 N95, R951 Ammonia, Methylamine Cartridge Standard 10/Pk.
18-947 N95, R951 Organic Vapor, Acid Gas, Ammonia, Methylamine, Formaldehyde, Hydrogen Fluoride Cartridge Standard 10/Pk.
19-003-215 P100 Particulate Cartridge Low-profile 10/Pk.
19-003-217 P100 Organic Vapor, Acid Gas Cartridge Standard 6/Pk.
19-003-222 P100 Chlorine, Mercury Vapor Cartridge Standard 6/Pk.
19-003-225 P100 Organic Vapor, Acid Gas Cartridge Short Stack 6/Pk.
19-063-985 N95, R951 Organic Vapor Cartridge Standard 10/Pk.
19-067-372 N95, R951 Organic Vapor, Acid Gas Cartridge Standard 10/Pk.
19-003-213 P100 Particulate Cartridge Standard 10/Pk.
19-003-223 P100 Organic Vapor Cartridge Short Stack 6/Pk.
19-003-219 P100 Organic Vapor, Acid Gas, Ammonia, Methylamine, Formaldehyde, Hydrogen Fluoride Cartridge Standard 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-003-216 Supplier MSA™ Supplier No. 815178
Add to Cart
Add to Cart
This item is not returnable. View return policy

Particulate, chemical, and combination cartridges for use with Comfo Classic™, Comfo Elite™, Ultra-Twin™ and Ultra Elite™ respirators

  • Low-profile particulate and ShortStack™ combination cartridges feature a patented low-profile design that improves field of vision
  • Special adapter also allows use with Advantage™ 200LS, Advantage 420, Advantage 4000 and Advantage 1000 respirators Sparkfoe™ particulate cartridge is suitable for use in applications with sparking hazard

Note: Do not exceed maximum use concentrations established by regulatory standards.

Certifications

All Comfo cartridges are approved under NIOSH 42CFR Part 84.

Specifications

Cartridge Type Organic Vapor Cartridge
Certifications/Compliance NIOSH 42CFR, Part 84
Color Black/Magenta
Design Type Standard
Filter Type P100
Product Type Respirator Cartridge
Filter Efficiency 99.97%
For Use With (Application) Asbestos abatement, painting, sanding, grinding, welding
For Use With (Equipment) Comfo classic™, ultra-twin™, ultra elite twin™ cartridge and duo-twin™ respirator and certain half mask and full face advantage™ respirator
Quantity 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.