Learn More
Description
- Low-profile particulate and ShortStack™ combination cartridges feature a patented low-profile design that improves field of vision
- Special adapter also allows use with Advantage™ 200LS, Advantage 420, Advantage 4000 and Advantage 1000 respirators Sparkfoe™ particulate cartridge is suitable for use in applications with sparking hazard
Note: Do not exceed maximum use concentrations established by regulatory standards.
Certifications
All Comfo cartridges are approved under NIOSH 42CFR Part 84.
Specifications
Specifications
| Cartridge Type | Organic Vapor, Acid Gas, Ammonia, Methylamine, Formaldehyde, Hydrogen Fluoride Cartridge |
| Certifications/Compliance | NIOSH 42CFR, Part 84 |
| Color | Olive/Magenta |
| Design Type | Standard |
| Filter Type | P100 |
| Product Type | Respirator Cartridge |
| Filter Efficiency | 99.97% |
| For Use With (Application) | Asbestos abatement, painting, sanding, grinding, welding |
| For Use With (Equipment) | Comfo classic™, ultra-twin™, ultra elite twin™ cartridge and duo-twin™ respirator and certain half mask and full face advantage™ respirator |
| Quantity | 6/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.