Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Comfo™ and Comfo Classic™ Half-Mask Respirators

Catalog No. 189992589B Shop All MSA Products
Change view
Click to view available options
Size:
Small
Medium
Large
Facepiece Material:
Hycar™
Silicone
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Facepiece Material
18-999-2589B Medium Hycar™
18-999-2588A Small Hycar™
18-999-2588B Medium Hycar™
18-999-2588C Large Hycar™
18-999-1511 Small Silicone
18-999-1510 Medium Silicone
18-999-1509 Large Silicone
18-999-2589A Small Hycar™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-999-2589B Supplier MSA™ Supplier No. 808053

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Requires

Comfo Series cartridges and filters (see separate listings).

Available in Hycar™ rubber or silicone, Comfo™ series half-mask respirators feature a unique face seal design for a comfortable fit.

  • Protection from exposure to vapors, gases, and particulates
  • SoftFeel™ facepiece material increases softness
  • Choose from Hycar™ rubber or silicone
  • Face seal design provides tight, comfortable fit
  • Ideal markets include utilities, oil & gas, construction and general industry

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Material Hycar™
Product Type Half-Mask Respirator
Size Medium
Facepiece Material Hycar™
For Use With (Application) Utilities, oil and gas, construction, general industry, welding
For Use With (Equipment) Comfo™ Series cartridges and filters
Harness Material Elastic
Model 808053
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.