Learn More
MSA™ Detector Tubes
Catalog No. 189993512 Shop All MSA Products
Change view
Click to view available options
Measurement Range:
1 to 50
Catalog No. Measurement Range
18-999-3512 1 to 50
1 options
Catalog No. 18-999-3512 Supplier MSA™ Supplier No. 804142
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Monitor air contaminants without extractions, analyses, or complicated calculations.
MSA/AUER detector tubes are a reliable method of testing for more than 120 hazardous gases and vapors. Detector tubes are flame sealed glass tubes containing treated adsorbent granules that react with a specific compound or group of compounds, causing the adsorbent to change color. A sample is collected by attaching the detector tube to a special bellows-type pump that draws a known volume of air with each stroke. After sampling, the length of adsorbent bed that has undergone the color change is measured from a scale printed on the tube, allowing direct reading of the vapor concentration – no extraction, no analysis, and no calculations are required.
Because they are simple to use, detector tubes have been used in a broad range of applications. They are particularly useful when quick screening is required, such as in Haz-Mat Response, and in industrial situations in which specific groups of compounds are monitored regularly.
Order Info
Designed for use with Kwik-Draw™ Pumps. Also can be used with Toximeter II Automatic Detector Tube pump, Dräger™ Model 31 Bellows Pump, and Dräger accuro™ Pump (see individual entries). Tubes come 10 per pack.
Specifications
Material Glass
Measurement Range 1 to 50
Quantity 10
Tip Type Break off