Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Evolution™ 5000 Thermal Imaging Camera Accessories

Catalog No. 19121425 Shop All MSA Products
Change view
Click to view available options
Product Type:
Battery Charger (110 V)
Battery Charger (220 V)
Caribiner
Carry Case
Lithium-ion Battery
Non-charging Mounting Bracket
Reflective Trim Kit
Retractable Lanyard
Shoulder Strap
Sun Shroud
Tripod Adapter
Wrist Strap
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-121-425 Battery Charger (220 V)
19-121-428 Wrist Strap
19-121-430 Retractable Lanyard
19-121-429 Shoulder Strap
19-121-431 Caribiner
19-121-424 Battery Charger (110 V)
19-121-426 Lithium-ion Battery
19-121-432 Sun Shroud
19-121-433 Reflective Trim Kit
19-121-437 Carry Case
19-121-435 Non-charging Mounting Bracket
19-121-436 Tripod Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 19-121-425 Supplier MSA™ Supplier No. 10040162

Specifications

Product Type Battery Charger (220 V)
For Use With (Equipment) Evolution™ 5000 Thermal Imaging Camera
Voltage 220 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.