Learn More
MSA™ Filter Covers for Advantage™ Cartridges
Catalog No. 19086975 Shop All MSA Products
Change view
Click to view available options
N95 Filter Cover (two required); 1 Ea.
Catalog No.
19-086975 N95 Filter Cover (two required); 1 Ea.
1 options
Catalog No. 19-086975 Supplier MSA™ Supplier No. 489353
Add to Cart
Add to Cart
For use with Advantage Cartridges
Specifications
Cartridge Type Respirator Cartridge
Filter Type N95
Product Type Snap-On Cover