Learn More
MSA™ FireHawk™ M7 Air Masks with PASS
Catalog No. 17200011 Shop All MSA Products
Click to view available options
Pressure:
High
Model:
Push-to-connect style regulator
Slide-to-connect style regulator
This item is not returnable. View return policy
Catalog No. 17-200-011
Supplier: MSA™ 10100092
This item is not returnable. View return policy
Includes
M-7 heads up display receiver, ipass
Ideal for industrial fire brigades
Designed to meet the needs of fire departments for basic, rugged and reliable SCBA without accessories.
  • Control module equipped with analog and digital pressure display for added redundancy
  • PASS icons display battery life, cylinder pressure, evacuation, thermal alarm and radio signal
  • Motion sensor alarm sounds at above 95dB
  • Datalogging of most recent 25 hours of use information
  • PR14™ first-stage regulator with 14 internal parts
  • Heads-up display and LED buddy lights
  • Stainless-steel cylinder band with secure cam-lock system
  • High-visibility reflective panels
  • Quick-Fill™ URC with Audi-Larm™ audible alarm
  • Firehawk™ MMR regulator
Order Info
Powered by four C cell batteries (not included). Facepiece and cylinder not included
Certifications
NIOSH 42 CFR Part 84 CBRN certified
Compliance
Meets NFPA 1981 and 1982-2007 Edition
Specifications
Certifications/Compliance NIOSH 42CFR, part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002
Product Type Air Mask
Pressure High
For Use With (Application) Protects against impact, water ingress, shields against chemical warfare agent contact, ideal for industrial fire brigades, confined space, firefighting, search and rescue
Includes M-7 heads-up display receiver, ipass
Model Slide-to-connect style regulator
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.