Learn More
MSA™ FireHawk™ M7 Air Masks with PASS
Shop All MSA Products
Click to view available options
Pressure:
High
Model:
Push-to-connect style regulator
Slide-to-connect style regulator
Description
Includes
M-7 heads up display receiver, ipass
Designed to meet the needs of fire departments for basic, rugged and reliable SCBA without accessories.
- Control module equipped with analog and digital pressure display for added redundancy
- PASS icons display battery life, cylinder pressure, evacuation, thermal alarm and radio signal
- Motion sensor alarm sounds at above 95dB
- Datalogging of most recent 25 hours of use information
- PR14™ first-stage regulator with 14 internal parts
- Heads-up display and LED buddy lights
- Stainless-steel cylinder band with secure cam-lock system
- High-visibility reflective panels
- Quick-Fill™ URC with Audi-Larm™ audible alarm
- Firehawk™ MMR regulator
Order Info
Powered by four C cell batteries (not included). Facepiece and cylinder not included
Certifications
NIOSH 42 CFR Part 84 CBRN certified
Compliance
Meets NFPA 1981 and 1982-2007 Edition
Specifications
Specifications
| Certifications/Compliance | NIOSH 42CFR, part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002 |
| Product Type | Air Mask |
| Pressure | High |
| For Use With (Application) | Protects against impact, water ingress, shields against chemical warfare agent contact, ideal for industrial fire brigades, confined space, firefighting, search and rescue |
| Includes | M-7 heads-up display receiver, ipass |
| Model | Push-to-connect style regulator |