Description
Basic, rugged and reliable SCBA without accessories.
- Choice of intermittent or continuous mode; user can switch among HUD modes
- Lower profile and easy-to-read pressure gauge
- Can be used with existing standalone PASS devices
- Upgrades easily to full PASS unit
- Stainless-steel cylinder band with secure cam-lock system
- High-visibility reflective panels
- Bell alarm
Order Info
Facepiece and cylinder not included
Compliance
Meets NFPA 1981 Edition
Specifications
Specifications
| Certifications/Compliance | NIOSH 42CFR, part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002 |
| Product Type | Air Mask |
| Pressure | Low |
| For Use With (Application) | Ideal for hazmat workers, first responders, firefighters, confined space, firefighting, search and rescue |
| For Use With (Equipment) | Standalone pass devices |
| Includes | Threaded hose and pressure gauge, CBRN solid cover |
| Model | Slide-to-connect with threaded hose and pressure gauge |