Description
Compliance
- Ballistic resistance: Meets NIJ 0108.01 IIIa standards
- Fragment/impact resistance: Meets Military ACH specifications
Specifications
Specifications
| Certifications/Compliance | NIJ 0108.01 IIIa |
| For Use With (Equipment) | ACH Advanced Combat Helmet |
| Product Type | Chinstrap Assembly |
| Quantity | 1 Ea. |
| Size | Universal |