Description
Compliance
- Ballistic resistance: Meets NIJ 0108.01 IIIa standards
- Fragment/impact resistance: Meets Military ACH specifications
Specifications
Specifications
| Certifications/Compliance | NIJ 0108.01 IIIa |
| For Use With (Equipment) | ACH Advanced Combat Helmet |
| Product Type | Pad Kit |
| Quantity | 1 Kit |
| Size | 8 |
| Thickness | 25.4 mm |