Learn More
MSA™ FP Vulcan™ Rescue Harnesses
Catalog No. 18039218 Shop All MSA Products
Change view
Click to view available options
X-Large
Catalog No.
18-039-218 X-Large
1 options
Catalog No. 18-039-218 Supplier MSA™ Supplier No. 10013654
  • Front, back, and shoulder D-rings provide attachment options
  • Compatible with SCBA back plate
  • Four sizes for superior fit and comfort
Ideal for emergency rescue teams, this is a confined-space and rescue harness rated for fall arrest. It features a side-entry vest-style harness with tongue-buckle leg straps. Front, back, and two shoulder D-rings provide alternative attachment options, with an adjustable sub-pelvic support strap. Comes with a front belay loop and attachment point for rescuing a disabled person. Attachment points also make this unit compatible with the back plates for the Ultralite™ 2, Ultralite MMR, and Custom 4500™ SCBAs (call your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information).
Compliance
Meet all requirements of UL/NFPA, ANSI A10.14, CSAZ259.10 and all applicable OSHA regulations.
Specifications
Size X-Large
D-Ring Location Back, Front, Shoulder
Certifications/Compliance Meets all requirements of UL/NFPA, ANSI A10.14, CSAZ259.10 and all applicable OSHA regulations.
For Use With (Application) Ideal for emergency rescue teams, this is a confined-space and rescue harness rated for fall arrest.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.