Learn More
MSA™ Gas Miser 4-Station Manifold
Catalog No. 189993339 Shop All MSA Products
Change view
Click to view available options
For use w/ Model RP and Econo-cal cylinders and Gas Miser regulator
Catalog No.
18-999-3339 For use w/ Model RP and Econo-cal cylinders and Gas Miser regulator
1 options
Catalog No. 18-999-3339 Supplier MSA™ Supplier No. 710274
Supply the correct amount of gas and bump test or calibrate up to four instruments simultaneously
The Gas Miser Manifold is not only a cylinder holder but it also incorporates a 4-station “bump test” or calibration manifold. It works with Model RP and Econo-Cal cylinders and Gas Miser regulator on any pumped or aspirated instrument. The manifold will supply the correct amount of gas required and will bump test or calibrate up to four instruments simultaneously. Gas Miser regulator sold separately.
Specifications
Product Type Manifold