Learn More
MSA™ Gas Miser™ Demand Regulator for RP Calibration Cylinders
Catalog No. 189993180 Shop All MSA Products
Change view
Click to view available options
Material:
Nickel Plated Brass/Polished Aluminum
Catalog No. Material
18-999-3180 Nickel Plated Brass/Polished Aluminum
1 options
Catalog No. 18-999-3180 Supplier MSA™ Supplier No. 710288
Add to Cart
Add to Cart
Supplies the exact gas flow for the connect instrument pump, removing the need to manually calibrate
  • Cost-efficient design
  • Turns itself off when not in use, simplifying the test and calibration process
  • Delivers test gas at a rate of .25lpm
  • Nickel-plated brass/polished aluminum
  • Polyurethane tubing
Specifications
Type Demand Regulator
Material Nickel Plated Brass/Polished Aluminum