Learn More
Description
- Color: Red
- Power: 12VDC
- Current Draw: 0.40 to 0.1 Amps
- Approval: NEMA 4X
Specifications
Specifications
| Color | Red |
| For Use With (Equipment) | Gas Monitor Remote Display |
| Type | Unit-mounted Strobe |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.