Learn More
MSA™ Goggle Retainers
Catalog No. 189993895 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
For MSA Caps
For MSA Hats
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-999-3895 For MSA Hats
19-041447 For MSA Caps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-3895 Supplier MSA™ Supplier No. 449895
Add to Cart
Add to Cart
Holds goggles securely on any MSA cap or hat
  • Sturdy neoprene-rubber accessory can be used with any MSA protective cap or hat
  • Retains goggle on helmet when not in use and holds goggle securely when worn for eye protection
Compliance
MSA goggle retainers are manufactured for use ONLY with MSA goggles in order to comply with ANSI/ISEA Z87.1-2010.
Specifications
Product Type Goggle Retainer
For Use With (Equipment) For MSA Hats
For Use With (Application) Holds goggles securely on any MSA cap or hat
Material Neoprene