Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Gravity™ Harness Class A, L, D

Catalog No. 19033901 Shop All MSA Products
Change view
Click to view available options
Gravity Harness Class A, L, D
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-033-901 Gravity Harness Class A, L, D
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-033-901 Supplier MSA™ Supplier No. SSH90925300

Suitable for controlled descent, positioning, ladder climbing, rescue, and fall arrest

Certifications

Meets ANSI Z359.1 and OSHA standards and is CSA certified. Certified according EN 361

Compliance

Meets ANSI™ Z359.1 and ANSI Z359.11-2014; ASTM™ F887-2013; CSA Z259.10; EN358:1999-12, EN361:2002 and EN813:2008 requirements.

Specifications

For Use With (Application) Controlled descent, positioning, ladder climbing, rescue, and fall arrest
Webbing Material Nylon
Buckle Style (Leg) Tongue
Certifications/Compliance Meets ANSI Z359.1 and OSHA standards and is CSA certified. Certified according EN 361
Capacity (Metric) 180 kg
Capacity (English) 400 lb.
Color Blue

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.