Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Jet-Eye Adapter for ALTAIR™ Multigas Detector

Catalog No. 18039325 Shop All MSA Products
Change view
Click to view available options
Length (English):
12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
18-039-325 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-039-325 Supplier MSA™ Supplier No. 10082834
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Jet-Eye Adapter with USB connector for ALTAIR multigas detectors

For use with ALTAIR multigas detectors

Specifications

Type Jet Eye IR Adapter
Length (English) 12 in.
Length (Metric) 30.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.