Learn More
MSA™ Line Trap Assembly
Catalog No. 18999107 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
260, 361 Gas Line
Catalog No. For Use With (Equipment)
18-999-107 260, 361 Gas Line
1 options
Catalog No. 18-999-107 Supplier MSA™ Supplier No. 74814
Add to Cart
Add to Cart
For sampling combustible gas in tanks, sewers or sumps
  • Prevents liquids from being drawn into instrument during sampling
  • Liquids trapped in removable plastic jar which mounts on case
Specifications
For Use With (Equipment) 260, 361 Gas Line
Type Line Trap Assembly