Description
- When multiple GALAXY™ System test stands are interconnected, data from all test stands is shared on a single memory card
- Industry-standard memory flash card available for simple data retention of more than 1000 individual test records
For use with MSA Galaxy Gas Detectors
Specifications
Specifications
| Type | Memory Card Reader |