Learn More
Description
- Accessory for Millennium or Advantage 1000 facepieces
- Polycarbonate outsert
- Available in clear or tinted
- Available in small or medium/large sizes
Specifications
Specifications
| Color | Tinted |
| Material | Polycarbonate |
| Product Type | Lens Outsert |
| Size | Small |
| For Use With (Application) | Provide additional impact protection |
| For Use With (Equipment) | Millennium™ or Advantage 1000 facepiece |
| Model | 10008909 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.