Learn More
MSA™ MiniCO™ Gas-Detection Responder Kits
Catalog No. 189993112 Shop All MSA Products
Change view
Click to view available options
Monitor with calibration gas, regulator and tubing
Monitor with jacket and strap, gas bump tester and CO detector tube cards
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-999-3112 Monitor with jacket and strap, gas bump tester and CO detector tube cards
19-034-599 Monitor with calibration gas, regulator and tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-3112 Supplier MSA™ Supplier No. 710551
19-034-599 includes monitor, calibration gas, regulator, and tubing; 18-999-3112 includes monitor with protective jacket and neck strap, Squirt™ Gas Bump Tester for in-field calibration, and 10 MiniCO carbon monoxide detector tube cards.