Learn More
Description
- Replacement black silicone nosecup
- Easily affixed to the facepiece without any tools
- Reduces the possibility of lens fogging
- Available in medium or large sizes
Specifications
Specifications
| Color | Black |
| Material | Silicone |
| Product Type | Nosecup Assembly |
| Size | Large |
| For Use With (Application) | Helps reduce lens fogging |
| For Use With (Equipment) | Ultravue and Ultra-Twin Full Facepieces |
| Model | 813140 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.