Learn More
MSA™ Passport™ PID II Organic Vapor Monitor Kit
Catalog No. 19042514 Shop All MSA Products
Change view
Click to view available options
Passport PID II II Organic Vapor Monitor kit
Catalog No.
19-042-514 Passport PID II II Organic Vapor Monitor kit
1 options
Catalog No. 19-042-514 Supplier MSA™ Supplier No. 711402
Includes
monitor, alkaline battery pack, pistol grip, 8 in. (20.3cm) probe, and a PVC carrying case.
  • Designed for hazardous material spills and underground tank leakage
  • Equally effective at emissions monitoring and general leak detection
Compact, portable instrument is designed to detect low concentrations of volatile organic compounds (VOCs) in a variety of monitoring applications. Though its primary use is for soil remediation associated with hazardous materials spills and underground tank leakage, it may also be used to monitor emissions and detect leaks. Passport PID II detects common compounds such as acetone, benzene, chlorobenzene, heptane, methyl tetrabutyl ether (MTBE), octane, phosphine, toluene, and many others.
Compliance
MET approved for use in Class I, Division 1, Groups A, B, C, and D; Class II, Division 2, Groups E, F, and G; Class III, Division 1 and 2 hazardous locations.