Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Passport™ VOC 2000 Organic Vapor and Oxygen Monitor Kit

Catalog No. 19042515 Shop All MSA Products
Change view
Click to view available options
Monitor with 10 ft. (3m) coiled PTFE sampling line
Monitor with 8 in. probe
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-042-515 Monitor with 10 ft. (3m) coiled PTFE sampling line
19-042-516 Monitor with 8 in. probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-042-515 Supplier MSA™ Supplier No. 10010864

Includes

VOC 2000 monitor, NiCd battery pack, 110V charger, 10 ft. (3m) coiled PTFE sampling line, 1 ft. (30cm) probe, steel belt clip, and standard packaging. VOC 2000 Monitor also available with NiCd battery pack, 110V charger, 8 in. (20.3cm) probe and pistol grip.

Operating range of 0 to 10,000ppm VOC and 0 to 100% O2

Photo-ionization allows this compact, portable instrument to provide very low-level leak detection of volatile organic compounds (VOCs). Perfect for chemical and petroleum industry applications, it detects low concentrations of compounds such as jet fuels, methyl ethyl ketone (MEK), benzene, and toluene. An integrated electrochemical oxygen sensor also monitors oxygen levels, making the monitor appropriate for many confined-space applications as well. Download data to computer via RS-232 cable.

Compliance

MET approved for use in Class I, Division 1, Groups A, B, C, and D; Class II, Division 2, Groups E, F, and G; Class III, Division 1 and 2 hazardous locations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.