Description
- Low-profile escape cylinder can be worn on either hip for user comfort
- Small size is less likely to snag when working in tight spaces
- Two attachments for keeping cylinder secure on belt
- Shoulder support strap incorporates hose keeper; regulator hose stays in place close to body
- Bracket/holder of second stage pressure regulator
- Nylon strap harness/carrier
Certifications
NIOSH approved as open-circuit, pressure-demand escape SCBA, open-circuit, pressure-demand escape SCBA and Type C supplied-air respirator
Compliance
NIOSH approved as open circuit; pressure-demand escape SCBA; open-circuit, pressure-demand escape SCBA; and Type C supplied-air respirator.
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Product Type | Cadet Escape Respirator |
| For Use With (Application) | Welding, chemical/petrochemical, hazardous materials, confined space entry, firefighting operation |
| Includes | 10 min. aluminum 3000 psi cylinder, nylon strap harness/carrier, ultra elite hycar™ facepiece with rubber head harness, hard case, first stage regulator and cylinder valve |
| Quantity | 1 Ea. |