Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Replacement Altair™ XCell™ Hydrogen Sulfide Sensor Kit

Catalog No. 17200170 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Altair XCell Multigas Detector
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-200-170 Altair XCell Multigas Detector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-200-170 Supplier MSA™ Supplier No. 10121213
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to detect and monitor low concentration hydrogen sulfide and carbon monoxide gas in air.

  • Long Life: >4-year life means fewer sensor replacements over the instrument's life
  • False-alarm Rejection: increased sensitivity means fewer false alarms, allowing you to work uninterrupted
  • Response: fast response time not only helps to save lives, but time and money as well

Specifications

Components Measured Hydrogen Sulfide
For Use With (Equipment) Altair XCell Multigas Detector
Type Replacement Hydrogen Sulfide Sensor Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.