Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Replacement Facepiece Assemblies for Ultra Elite™ and Ultravue™ SCBAs

Catalog No. 19005402C Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Firehawk™ MMR Air Mask
Ultralite MMR and custom 4500 MMR air mask
Material:
Hycar™ Rubber
Silicone
Size:
Small
Medium
Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material Size
19-005-402C Firehawk™ MMR Air Mask Hycar™ Rubber Large
19-815-702 Ultralite MMR and custom 4500 MMR air mask Silicone Large
18-999-3166 Ultralite MMR and custom 4500 MMR air mask Hycar™ Rubber Large
18-999-3153 Ultralite MMR and custom 4500 MMR air mask Silicone Medium
18-999-3168 Ultralite MMR and custom 4500 MMR air mask Hycar™ Rubber Small
18-999-3162 Ultralite MMR and custom 4500 MMR air mask Hycar™ Rubber Medium
19-005-402B Firehawk™ MMR Air Mask Hycar™ Rubber Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-005-402C Supplier MSA™ Supplier No. 10024431
Add to Cart
Add to Cart

Specifications

Certifications/Compliance NIOSH 42CFR, part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002
Color Black
Product Type MMR Facepiece
For Use With (Application) For monitoring air cylinder contents, firefighting
For Use With (Equipment) Firehawk™ MMR Air Mask
Includes Speed-on™ head harness, nosecup
Material Hycar™ Rubber
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.