Learn More
Description
Convenient, belt-mounted pouch with Velcro closures. Made of easy-to-clean vinyl, it is designed to carry half-mask respirators. Gray.
Specifications
Specifications
| Product Type | Respirator Carrying Pouch |
| Closure Type | Velcro™ |
| Color | Gray |
| Material | Vinyl |
| Model | 468730 |
| Includes | Convenient, Belt-mounted Pouch |
| For Use With (Equipment) | Half-mask Respirator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.