Learn More
MSA™ Safe Escape CBRN Respirators
Designed to escape sudden chemical, biological, radiological, or nuclear hazards
Supplier:  MSA™ 10066792
 View more versions of this product
Catalog No. 19-161-113
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Translucent hood with one-piece large bonded lens offers secure protection and a wide field of view
  • Canister is permanently bonded to the respirator for a secure connection
  • Rubber neck seal provides a comfortable and reliable seal
  • Elastic hair tie allows for the securing of hair before donning the respirator
  • Nose cup helps reduce lens fogging and carbon dioxide buildup
  • Hexagonal storage case is strong, lightweight, and easily accessible
  • 15 minute air supply
Standard
  • Color-coded labels on respirator
  • Tactical
    • Black/gray labels on respirator
    Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
    Certifications
    NIOSH - 10052317
    Specifications
    Specifications
    NIOSH
    15 min.
    CBRN incidents, civil disturbances, clandestine labs
    10066792
    Standard Escape Respirator
    15 min.
    Bonded lens and cbrn canister, high stretch rubber neck seal, nose cup
    Large
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Promotions
    Promotions
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.