Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSA™ Safe Escape CBRN Respirators
Designed to escape sudden chemical, biological, radiological, or nuclear hazards
Specifications
Certifications/Compliance | NIOSH |
---|---|
Duration | 15 min. |
Cycle Time | 15 min. |
For Use With (Application) | CBRN incidents, civil disturbances, clandestine labs |
Includes | Bonded lens and cbrn canister, high stretch rubber neck seal, nose cup |
Catalog Number | Mfr. No. | Product Type | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Size | Price | Quantity & Availability | ||||
19-161-114
|
MSA™
10066787 |
Tactical Escape Respirator | Small | N/A | N/A | N/A | |||
19-161-115
|
MSA™
10066788 |
Tactical Escape Respirator | Medium | N/A | N/A | N/A | |||
19-161-116
|
MSA™
10066789 |
Tactical Escape Respirator | Large | N/A | N/A | N/A | |||
19-161-113
|
MSA™
10066792 |
Standard Escape Respirator | Large | N/A | N/A | N/A | |||
19-161-112
|
MSA™
10066791 |
Standard Escape Respirator | Medium | N/A | N/A | N/A | |||
19-161-111
|
MSA™
10066790 |
Standard Escape Respirator | Small | N/A | N/A | N/A | |||
Description
- Translucent hood with one-piece large bonded lens offers secure protection and a wide field of view
- Canister is permanently bonded to the respirator for a secure connection
- Rubber neck seal provides a comfortable and reliable seal
- Elastic hair tie allows for the securing of hair before donning the respirator
- Nose cup helps reduce lens fogging and carbon dioxide buildup
- Hexagonal storage case is strong, lightweight, and easily accessible
- 15 minute air supply
Standard
- Color-coded labels on respirator
- Black/gray labels on respirator
Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
Certifications
NIOSH - 10052317
Specifications
NIOSH | |
15 min. | |
Bonded lens and cbrn canister, high stretch rubber neck seal, nose cup |
15 min. | |
CBRN incidents, civil disturbances, clandestine labs |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title