Description
Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
Certifications
NIOSH - 10052317
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Product Type | Training Hood |
| For Use With (Application) | CBRN incidents, civil disturbances, clandestine labs |
| For Use With (Equipment) | Emergency escape respirator |
| Model | 10066785 |
| Size | Medium |