MSA™ Sirius™ Multigas Detector Calibration Gas Cylinders
For use with Sirius Multigas Detectors.
Manufacturer: MSA™ 10048981
 View more versions of this product
Catalog No. 19-123-452
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Sirius Multigas detectors
    Capacity (Metric) 58L
    Includes 2.5% CH4, 15% O2, 60ppm CO, 20ppm H2S
    Product Type Econo-Cal CH4 cylinder
    Capacity 1961.2 oz., 58L
    Capacity (English) 15.3 gal.
    For use with Sirius Multigas Detectors.
  • Description & Specifications
Related Products
Product Content Feedback