Learn More
Description
Includes
Economy Models include: Monitor, sensors and batteries
Integrates high-performance photoionization detector sensor with a multigas detector for up to four gases. Users are able to simultaneously monitor for volatile organic compounds (VOCs) with low vapor pressures while measuring for combustible, toxic, and oxygen-deficient atmospheres within one reliable, easy-to-use, durable unit.
Features:
• Measures VOC readings first for greater accuracy of ±10% or 2ppm over full range
• Best in class humidity performance
• Notifies failure or removal of lamp or ion chamber
• Features an over range indicator, external exhaust, and multidirectional audio and visual alarms
• Lamps are easy to replace and clean
For use in:
• Leak detection
• Perimeter monitor
• Spill delineation
• Remediation
• Arson investigation
• Diesel fuel TVL monitoring (Marine Chemist)
• Decontamination
• PPE Determination
Compliance
Meets 47CFR part 15. UL913 6th edition and CSA 22.2 No. 157 for use in hazardous locations Class I, Divisions I, Groups A, B, C, and D. Temperature class T3C. Meets Canadian combustible performance standard CSA 22.2 No. 152 for methane only.
Specifications
Specifications
| Sensors | LEL, O2, CO |
| Sensor Range | 0 to 100% LEL, 0 to 25% Vol O2, 0 to 500 ppm CO |
| Certifications/Compliance | 47CFR, UL913, CSA 22.2, T3C |
| Depth (Metric) | 6.65 cm |
| Depth (English) | 2.62 in. |
| Includes | Monitor, Sensors and Batteries |
| Temperature (Metric) Operating | 0° to 40°C |
| Width (Metric) | 9.19 cm |
| Accuracy | ±10% |
| Height (Metric) | 16.51 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.