Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSA™ Sirius™ Multigas Detector - Economy Model
Combines PID functionality with a four-gas monitor
Specifications
| Accuracy | ±10% |
|---|---|
| Certifications/Compliance | 47CFR, UL913, CSA 22.2, T3C |
| Depth (English) | 2.62 in. |
| Depth (Metric) | 6.65 cm |
| For Use With (Application) | Leak Detection, Perimeter monitor, Spill delineation, Remediation, Arson investigation, Diesel fuel TVL monitoring, Decontamination, PPE determination |
Includes: Economy Models include: Monitor, sensors and batteries
| Catalog Number | Mfr. No. | Sensors | Model | Sensor Range | Sensor Resolution | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sensors | Model | Sensor Range | Sensor Resolution | Price | Quantity & Availability | ||
|
19-123-408
|
MSA™
10051151 |
LEL, O2, CO, H2S | 0 to 100% LEL, 0 to 25% Vol O2, 0 to 500 ppm CO, 0 to 200 ppm H2S | 1% LEL, 0.1% Vol O2, 1 ppm CO, 1 ppm H2S | N/A | N/A | N/A | ||
|
19-123-409
|
MSA™
10051152 |
0 to 100% LEL, 0 to 25% Vol O2, 0 to 200 ppm H2S | N/A | N/A | N/A | ||||
|
19-123-410
|
MSA™
10051153 |
LEL, O2, CO | 0 to 100% LEL, 0 to 25% Vol O2, 0 to 500 ppm CO | N/A | N/A | N/A | |||
|
19-123-411
|
MSA™
10051154 |
LEL, O2, CO, H2S | 0 to 100% LEL, 0 to 25% Vol O2, 0 to 500 ppm CO, 0 to 200 ppm H2S | 1% LEL, 0.1% Vol O2, 1 ppm CO, 1 ppm H2S | N/A | N/A | N/A | ||
|
19-123-421
|
MSA™
10051186 |
N/A | N/A | N/A | |||||
|
19-123-420
|
MSA™
10051185 |
CH4, O2, CO, H2S | CH4, 0 to 25% Vol O2, 0 to 500 ppm CO, 0 to 200 ppm H2S | CH4, 0.1% Vol O2, 1 ppm CO, 1 ppm H2S | N/A | N/A | N/A | ||
|
19-123-422
|
MSA™
10051187 |
N/A | N/A | N/A | |||||
|
19-123-423
|
MSA™
10051188 |
N/A | N/A | N/A | |||||
Description
Integrates high-performance photoionization detector sensor with a multigas detector for up to four gases. Users are able to simultaneously monitor for volatile organic compounds (VOCs) with low vapor pressures while measuring for combustible, toxic, and oxygen-deficient atmospheres within one reliable, easy-to-use, durable unit.Features:
- Measures VOC readings first for greater accuracy of ±10% or 2ppm over full range
- Best in class humidity performance
- Notifies failure or removal of lamp or ion chamber
- Features an over range indicator, external exhaust, and multidirectional audio and visual alarms
- Lamps are easy to replace and clean
For use in:
- Leak detection
- Perimeter monitor
- Spill delineation
- Remediation
- Arson investigation
- Diesel fuel TVL monitoring (Marine Chemist)
- Decontamination
- PPE Determination
Compliance
Meets 47CFR part 15. UL913 6th edition and CSA 22.2 No. 157 for use in hazardous locations Class I, Divisions I, Groups A, B, C, and D. Temperature class T3C. Meets Canadian combustible performance standard CSA 22.2 No. 152 for methane only.
Specifications
| ±10% | |
| 2.62 in. | |
| Leak Detection, Perimeter monitor, Spill delineation, Remediation, Arson investigation, Diesel fuel TVL monitoring, Decontamination, PPE determination | |
| 6.5 in. | |
| Monitor, Sensors and Batteries | |
| 0° to 40°C |
| 47CFR, UL913, CSA 22.2, T3C | |
| 6.65 cm | |
| Multigas Detector Economy Kit | |
| 16.51 cm | |
| 32° to 104°F | |
| 9.19 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title