Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Sirius™ Multigas Detector Rubber Boot

Catalog No. 19123437 Shop All MSA Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-123-437 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-123-437 Supplier MSA™ Supplier No. 10052514

Rubber boot for Sirius multigas detectors

For use with Sirius multigas detectors

  • Color: Black
  • Comes with harness

Specifications

Color Black
Type Rubber Boot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.