Learn More
MSA™ TechnaCurv™ Construction Harnesses
Catalog No. 191303873 Shop All MSA Products
Change view
Click to view available options
Size:
Standard
Super X-Large
X-Large
X-Small
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-3873 Super X-Large
19-130-3870 X-Small
19-130-3871 Standard
19-130-3872 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-130-3873 Supplier MSA™ Supplier No. 10063657
Specifications
Certifications/Compliance ANSI Z359.1, ANSI A10.32, CSA Z259.10, OSHA
Buckle Style (Chest) Qwik-Fit™
Product Type Vest-style Harness
Size Super X-Large
Buckle Style (Leg) Secure-Fit™ tongue leg strap buckle
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.