Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ TechnaCurv™ Full-Body Vest-Style Harnesses

Catalog No. 17200103 Shop All MSA Products
Change view
Click to view available options
Buckle Style (Leg):
Tongue
D-Ring Location:
Back
Back, Chest, Hips
Includes:
Qwik-fit chest strap, manual
Size:
Standard
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style (Leg) D-Ring Location Includes Size
17-200-103 Tongue Back Qwik-fit chest strap, manual X-Large
17-200-108 Tongue Back, Chest, Hips Qwik-fit chest strap, manual Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-200-103 Supplier MSA™ Supplier No. 10041592

missing translation for 'validMainframeMsg'

Made of soft, flexible webbing

Optional Secure-Fit leg buckles with positive engagement windows allow you to see buckle is properly engaged—no need to listen for a click on a noisy site.

  • Soft, flexible curvilinear torso webbing moves with the wearer for improved comfort and a secure fit
  • Webbing curves to fit around the neck, so it will not chafe
  • Lightweight polymer Y-back D-locator pad allows complete adjustment of back D-ring for proper fit
  • Durable visco-elastic shoulder and sub-pelvic pads permit air to circulate; keep wearer cool and comfortable
  • Single-hand operation for easy donning and doffing
  • Feature a standard Qwik-Fit™ chest strap

Order Info

Maximum Capacity: 400 lb. (181kg).

Compliance

Meets applicable OSHA standards; ANSI Z359.11-2014: CSA Z259.10-12 requirements.

Specifications

Color Red
Size X-Large
Includes Qwik-fit chest strap, manual
Style Vest
Webbing Material Teflon-coated Polyester
Product Type Full-body harness
Certifications/Compliance OSHA, ANSI Z359.1 and A10.32, CSA Z259.10. Certified according to EN 361and EN 358
Buckle Style (Leg) Tongue
D-Ring Location Back
Buckle Style Tongue
Capacity (English) 400 lb.
For Use With (Application) Confined space, construction
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.