Learn More
MSA™ Technacurv™ Tower Full-Body Harnesses
Curves webbing around the neck area to help prevent chafing and discomfort
Supplier: MSA™ 10048345
View more versions of this product
Description
- Variable width webbing in sub-pelvic area distributes weight over a wider area for increased comfort and support
- PTFE-coated webbing resists water, oil, grease and dirt
- Adjustable D-rings
- Qwik-Fit chest buckle
- Integral body belt with two side D-rings and two accessory rings and removable saddle with adjustable height
- Secure-Fit buckles feature single-hand operation, and positive engagement window
- Adjustable Y-back D-locator pad allows complete adjustment of back D-ring for proper fit
Certifications
CSA certified
Compliance
Meet applicable OSHA and ANSI standards.
Specifications
Back, Chest, Hips | |
CSA certified; Meet applicable OSHA and ANSI standards. | |
Vest |
X-Large | |
Secure-Fit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.