Description
This slotted cap is intended for use where workers may need a helmet with elevated temperature capability.
- Injection-molded, rivetless shell of glass-reinforced nylon has beaded edge for improved rigidity
- Lightweight, high-strength helmets for use in elevated temperatures; withstands radiant heat loads up to, but not exceeding, 350°F
- Slots in cap allow for mounting of MSA hearing protection, welding helmets, and V-Gard System face protection products, as well as other MSA helmet accessories
Compliance
Third-party certified by SEI to meet the Type I, Class G requirements of ANSI/ISEA Z98.1 and CSA Z94.1.
Specifications
Specifications
| Certifications/Compliance | Meets ANSI Z89.1-2003; Type I; Class G and CSA Z94.1-2005 (Class G). |
| Color | Green |
| Suspension Type | Fas-Trac Ratchet |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.