Description
Includes
Complete assembly includes two Window-Cator canisters (one required for operation), facepiece, breathing tube, harness, and carrying case.
Offers P100 protection against a variety of contaminants; Unique Window-Cator™ canister enables wearer to see when replacement is necessary.
Mask provides respiratory protection against acid gases, organic vapors, carbon monoxide, and ammonia.
Maximum Use Conditions for Type N gas mask: Entry, 100 times the Permissible Exposure Level without exceeding the Immediately Dangerous to Life or Health (IDLH) concentration; Escape, contaminant concentrations up to 20,000ppm (2% by volume) acid gases, carbon monoxide, and organic vapors, 30,000ppm (3%) ammonia, or 2% total when more than one class is present. Also effective against hydrogen fluoride gas.
Medium-sized facepiece is constructed of black Hycar™ rubber with wraparound polycarbonate lens for minimal visual distortion. Features include inhalation check valve that prevents exhaled air from entering breathing tube, mechanical speaking diaphragm for short-range communication, and five adjustable rubber straps and a vinyl neck strap for holding facepiece in standby position.
Certifications
Canister has P100 filter type and efficiency rating and is NIOSH approved.
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Product Type | Gas Mask Assembly |
| For Use With (Application) | Respiratory protection for acids gas, organic vapors, carbon monoxide and ammonia |
| Includes | 2 Window-Cator canisters (one required for operation), facepiece, breathing tube, harness, and carrying case. |