Description
Includes
Speed-on™ head harness, nosecup, firehawk™ m7 heads up display bracket, standard speaking diaphragm
Provides enhanced peripheral and downward vision
- Resists chemical attack, temperature extremes, and rugged use without tearing
- Seals securely with wide range of facial contours
- Five adjustable straps with stainless-steel, field-replaceable buckles for comfortable fit
- Standard-speaking diaphragm
- Rugged polycarbonate lens with scratch-resistant coating
- Choice of push-to-connect or slide-to-connect style regulators
Specifications
Specifications
| Certifications/Compliance | NFPA 2007 |
| Product Type | Facepiece Assembly |
| For Use With (Application) | Resists chemical attack, temperature extremes, rugged use without tearing |
| For Use With (Equipment) | Firehawk™ m7 air mask |
| Includes | Speed-on™ head harness, nosecup, Firehawk™ M7 heads-up display bracket, standard speaking diaphragm |
| Size | Medium |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.