Learn More
MSA™ Ultra Elite™ M7 Facepiece
Catalog No. 17200020 Shop All MSA Products
Change view
Click to view available options
Product Type:
Facepiece Assembly
M-7 Facepiece
Size:
Small
Medium
Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Size
17-200-020 M-7 Facepiece Medium
17-200-018 M-7 Facepiece Small
17-200-022 M-7 Facepiece Large
17-200-017 Facepiece Assembly Small
17-200-019 Facepiece Assembly Medium
17-200-021 Facepiece Assembly Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-200-020 Supplier MSA™ Supplier No. 10084824
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Speed-on™ head harness, nosecup, firehawk™ m7 heads up display bracket, standard speaking diaphragm
For use with FireHawk™ M7 air masks
Provides enhanced peripheral and downward vision
  • Resists chemical attack, temperature extremes, and rugged use without tearing
  • Seals securely with wide range of facial contours
  • Five adjustable straps with stainless-steel, field-replaceable buckles for comfortable fit
  • Standard-speaking diaphragm
  • Rugged polycarbonate lens with scratch-resistant coating
  • Choice of push-to-connect or slide-to-connect style regulators
Specifications
Certifications/Compliance NFPA 2007
Product Type M-7 Facepiece
For Use With (Application) Resists chemical attack, temperature extremes, rugged use without tearing
For Use With (Equipment) Firehawk™ m7 air mask
Includes Speed-on™ head harness, nosecup, Firehawk™ M7 heads-up display bracket, standard speaking diaphragm
Size Medium
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.