Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Ultra-Twin™ Full Facepiece Respirators

Catalog No. 18916 Shop All MSA Products
Change view
Click to view available options
Size:
Small
Medium
Large
Facepiece Material:
Hycar™
Silicone
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Facepiece Material
18-916 Small Silicone
18-999-3307 Small Hycar™
18-619W Medium Hycar™
18-619X Large Hycar™
18-922 Medium Silicone
18-917 Large Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-916 Supplier MSA™ Supplier No. 480263
Add to Cart
Add to Cart

Requires

Comfo Series cartridges and filters (see separate listings).

Respirators provide a secure fit and strong protection while offering several options including an inturned lip for a comfortable seal against the face and a large chin cup for easy positioning.

  • Five suspension headstraps and roller buckles secure the facepiece while minimizing hair entanglement
  • Wide coated polycarbonate lens resists scratching and chemicals
  • Available in Hycar™™ rubber and silicone

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Threaded
Color Black
Material Silicone
Product Type Full Facepiece Respirator
Size Small
Facepiece Material Silicone
For Use With (Application) Utilities, general industry, welding
For Use With (Equipment) Comfo™ Series cartridges and filters
Includes Nosecup and speaking diaphragm
Lens Type Anti-scratch
Material (Lens) Polycarbonate
Model 480263
Suspension Type 5-point
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.