Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Ultra-Twin™ Full Facepiece Respirators

Respirators provide a secure fit and strong protection while offering several options including an inturned lip for a comfortable seal against the face and a large chin cup for easy positioning.

$866.88 - $907.96

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Threaded
Color Black
Product Type Full Facepiece Respirator
For Use With (Application) Utilities, general industry, welding
View More Specs

Requires: Comfo Series cartridges and filters (see separate listings).


Products 6
Catalog Number Mfr. No. Size Facepiece Material Price Quantity & Availability  
Catalog Number Mfr. No. Size Facepiece Material Price Quantity & Availability  
18-999-3307
Accessories View Documents
MSA™
471298
Small Hycar™
Each for $866.88
Only null left
Add to Cart
 
18-619W
Accessories View Documents
MSA™
471286
Medium Hycar™
Each for $866.88
Only null left
Add to Cart
 
18-619X
Accessories View Documents
MSA™
471310
Large Hycar™
Each for $866.88
Only null left
Add to Cart
 
18-916
Accessories View Documents
MSA™
480263
Small Silicone
Each for $866.88
Only null left
Add to Cart
 
18-922
Accessories View Documents
MSA™
480259
Medium Silicone
Each for $907.96
Only null left
Add to Cart
 
18-917
Accessories View Documents
MSA™
480267
Large Silicone
Each for $907.96
Only null left
Add to Cart
 
Description

Description

  • Five suspension headstraps and roller buckles secure the facepiece while minimizing hair entanglement
  • Wide coated polycarbonate lens resists scratching and chemicals
  • Available in Hycar™™ rubber and silicone

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Specifications

Specifications

NIOSH 42 CFR Part 84
Black
Utilities, general industry, welding
Nosecup and speaking diaphragm
Polycarbonate
Threaded
Full Facepiece Respirator
Comfo™ Series cartridges and filters
Anti-scratch
5-point
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.