Learn More
MSA™ V-Gard™ 500 Vented Protective Cap
Catalog No. 19005348 Shop All MSA Products
Change view
Click to view available options
Color:
Blue
Green
Hi-vis Orange
Red
White
Suspension Type:
4-Point Ratchet
6-Point Ratchet
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-005-348 Blue 6-Point Ratchet
19-005-351 Red 4-Point Ratchet
19-005-346 White 6-Point Ratchet
19-005-345 White 4-Point Ratchet
19-005-353 Green 4-Point Ratchet
19-005-355 Hi-vis Orange 4-Point Ratchet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-005-348 Supplier MSA™ Supplier No. 10034028
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cooling vents along the crown improve air circulation and comfort
  • Comfortable, lightweight design
  • Polyethylene shell features contours above the ear for easy integration of hearing protection
  • Glaregard™ underbrim reduces reflective glare
  • Available with four- or six-point Fas-Trac™ ratchet suspension with absorbent sweatband
Compliance
Third-party certified to meet ANSI/ISEA Z98.1, Type I, Class C requirements.
Specifications
Certifications/Compliance Meets or exceeds ANSI Z89.1-2003, Class C and CSA Z94.1-2005.
Color Blue
Suspension Type 6-Point Ratchet
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.