Learn More
MSA™ V-Gard™ Protective Hat
Catalog No. 19815603 Shop All MSA Products
Change view
Click to view available options
Color:
Gold
Hi-vis Orange
Orange
Includes:
Fast-Trac suspension
Ratchet suspension
Staz-On supsoension
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
19-815-603 Orange Fast-Trac suspension
18-999-3959 Hi-vis Orange Staz-On supsoension
18-999-3890 Gold Ratchet suspension
19-815-630 Hi-vis Orange Fast-Trac suspension
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-815-603 Supplier MSA™ Supplier No. 496075
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Certifications/Compliance ANSI/ISEA Z89.1-2009, CSA Z94.1-2005
Color Orange
Includes Fast-Trac suspension
Material Polyethylene
Product Type Hard Hat
Quantity 1 Ea.
Size (Metric) 165.1 to 203.2 mm
Suspension Type 4-point Webbed Fas-Trac™ Ratchet
Head Style Full Brim (Slotted)
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.