Learn More
MSA™ V-Gard™ Replacement Steel Mesh Visors
Provide maximum ventilation in humid working conditions
$17.25 - $17.77
Specifications
For Use With (Equipment) | V-Gard Frame and Geadgear |
---|
Alert:
Do not use where splash, elevated temperatures, UV or impact hazards exist
Catalog Number | Mfr. No. | dimProductType | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | dimProductType | Price | Quantity & Availability | |||||
17-200-039
|
MSA™
10116557 |
Each for $17.77
|
|
||||||
17-200-040
|
MSA™
10116558 |
Each for $17.25
|
|
||||||
Description
- Lightweight and durable — great for outdoor applications
- Epoxy-coated 20 x 20 mesh bound with heavy-duty plastic
- Nitrometer style can be used only with V-Gard retractable of standard chin protector
Compliance
Meet EN 1731-F, ANSI/ISEA Z87.1-2010 (general protector) and AS/NZS 1337
Specifications
V-Gard Frame and Geadgear |